Merck
  • A Double-Negative Feedback Interaction between MicroRNA-29b and DNMT3A/3B Contributes to Ovarian Cancer Progression.
A Double-Negative Feedback Interaction between MicroRNA-29b and DNMT3A/3B Contributes to Ovarian Cancer Progression.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2016-11-11)
Yue Teng, Xiaohang Zuo, Meng Hou, Yan Zhang, Chen Li, Wenjuan Luo, Xu Li
ABSTRACT
Epigenetic abnormalities are increasingly observed in multiple malignancies, including epithelial ovarian cancer (EOC), and their effects can be significantly counteracted by tumor-suppressor microRNAs, namely epi-miRNAs. Here, we investigated the role of miR-29b, a well-established epi-miRNA, in the DNA methylation regulation of EOC cells. The correlation between miR-29b and DNMT3A/3B expression was evaluated by RT-qPCR, western blotting and immunohistochemical analysis. The functional roles of miR-29b and DNMT3A/3B were tested by anti-miRs and microRNA precursors. A luciferase reporter assay was employed to detect the direct binding of miR-29b to DNMT3A/3B 3' UTRs. Co-IP was utilized for investigating Id-1 binding activity. miR-29b was negatively correlated with DNMT3A/3B expression at the cellular/histological levels. miR-29b silencing was correlated with increased DNMT3A/3B levels, whereasmiR-29b over-expression caused DNMT3A/3B down-regulation. Luciferase reporter assays confirmed that the miR-29b-mediated downregulation of DNMT3A/3Boccurred through the direct targeting of theirmRNAs'3'-UTRs,whereasBGS assays found that DNMT3A/3B knockdown increased miR-29b expression via CpG island promoter hypomethylation, thus suggesting a crucial crosstalk betweenmiR-29b and DNMT3A/3B via a double-negative feedback loop. Co-IP assay confirmed direct binding between DNMT3A and Id-1. Taken together, our study sheds light on a novel epigenetic circuitry regulating EOC progression and may provide novel options for miR-29b-based epi-therapeutic approaches for future EOC treatment.