- Endoplasmic Reticulum (ER) Stress Induces Sirtuin 1 (SIRT1) Expression via the PI3K-Akt-GSK3β Signaling Pathway and Promotes Hepatocellular Injury.
Endoplasmic Reticulum (ER) Stress Induces Sirtuin 1 (SIRT1) Expression via the PI3K-Akt-GSK3β Signaling Pathway and Promotes Hepatocellular Injury.
Sirtuin 1 (SIRT1), an NAD(+)-dependent histone deacetylase, plays crucial roles in various biological processes including longevity, stress response, and cell survival. Endoplasmic reticulum (ER) stress is caused by dysfunction of ER homeostasis and exacerbates various diseases including diabetes, fatty liver, and chronic obstructive pulmonary disease. Although several reports have shown that SIRT1 negatively regulates ER stress and ER stress-induced responses in vitro and in vivo, the effect of ER stress on SIRT1 is less explored. In this study, we showed that ER stress induced SIRT1 expression in vitro and in vivo. We further determined the molecular mechanisms of how ER stress induces SIRT1 expression. Surprisingly, the conventional ER stress-activated transcription factors XBP1, ATF4, and ATF6 seem to be dispensable for SIRT1 induction. Based on inhibitor screening experiments with SIRT1 promoter, we found that the PI3K-Akt-GSK3β signaling pathway is required for SIRT1 induction by ER stress. Moreover, we showed that pharmacological inhibition of SIRT1 by EX527 inhibited the ER stress-induced cellular death in vitro and severe hepatocellular injury in vivo, indicating a detrimental role of SIRT1 in ER stress-induced damage responses. Collectively, these data suggest that SIRT1 expression is up-regulated by ER stress and contributes to ER stress-induced cellular damage.