Merck
Chronic exposure to K
Neuropharmacology (2016-09-14)
Elizabeth Haythorne, D Lee Hamilton, John A Findlay, Craig Beall, Rory J McCrimmon, Michael L J Ashford
ABSTRACT
Individuals with Type 1 diabetes (T1D) are often exposed to recurrent episodes of hypoglycaemia. This reduces hormonal and behavioural responses that normally counteract low glucose in order to maintain glucose homeostasis, with altered responsiveness of glucose sensing hypothalamic neurons implicated. Although the molecular mechanisms are unknown, pharmacological studies implicate hypothalamic ATP-sensitive potassium channel (K
Sigma-Aldrich
NN414, ≥98% (HPLC)