Merck
  • Dynamic kinetic resolution of primary amines with a recyclable Pd nanocatalyst for racemization.
Dynamic kinetic resolution of primary amines with a recyclable Pd nanocatalyst for racemization.
Organic letters (2007-02-13)
Mahn-Joo Kim, Won-Hee Kim, Kiwon Han, Yoon Kyung Choi, Jaiwook Park
ABSTRACT
A practical procedure for the dynamic kinetic resolution (DKR) of primary amines has been developed. This procedure employs a palladium nanocatalyst as the racemization catalyst, a commercial lipase (Novozym-435) as the resolution catalyst, and ethyl acetate or ethyl methoxyacetate as the acyl donor. Eleven primary amines and one amino acid amide have been efficiently resolved with good yields (85-99%) and high enantiomeric excesses (97-99%). [reaction: see text]
Sigma-Aldrich
Palladium nanoparticles entrapped in aluminum hydroxide matrix, 0.5 wt. % loading