- A case of etizolam dependence.
A case of etizolam dependence.
Indian journal of pharmacology (2014-12-30)
Sumit Gupta, Bhavuk Garg
PMID25538342
ABSTRACT
Etizolam is a thienodiazepine anxiolytic which is said to have lower dependence potential than other benzodiazepines. We report a case of etizolam dependence in a young male with social anxiety disorder and moderate depression. This case report highlights the fact that the same caution be exercised while prescribing etizolam with respect to its potential to cause dependence as with any other benzodiazepine.
Product Description
Supelco
Etizolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®