Merck
  • Comparative transcriptomics and comprehensive marker resource development in mulberry.
Comparative transcriptomics and comprehensive marker resource development in mulberry.
BMC genomics (2016-02-06)
Bushra Saeed, Vinay K Baranwal, Paramjit Khurana
ABSTRACT
High potential of Morus laevigata and Morus serrata has been proposed in the breeding programs for Morus sp. However, due to the lack of dense molecular markers this goal is still in its nascent stage and not yet realized. We thus, sequenced the transcriptomes of these two wild Morus species and utilized the data for marker development. We generated 87.0 and 80.3 Mb of transcriptome data from M. laevigata and M. serrata, respectively. The transcriptomes from M. laevigata and M. serrata, were assembled into 95,181 and 85,269 transcripts, respectively, and annotated. We identified around 24,049 Simple Sequence Repeats (SSRs), 1,201,326 Single Nucleotide Polymorphisms (SNPs) and 67,875 Insertion-Deletions (InDels). The variants having a higher impact were also identified and their effect was further investigated. The transcriptome resource from the wildly growing mulberry species developed in this study can find wide applicability in gene identification and/or characterization. It can also contribute immensely in the existing mulberry improvement programs.
Product Description
Supelco
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate, for chiral derivatization, LiChropur, ≥98.0% (HPLC)