Merck
  • Concise synthesis of enantiopure alpha-trifluoromethyl alanines, diamines, and amino alcohols via the Strecker-type reaction.
Concise synthesis of enantiopure alpha-trifluoromethyl alanines, diamines, and amino alcohols via the Strecker-type reaction.
The Journal of organic chemistry (2006-08-26)
Florent Huguenot, Thierry Brigaud
ABSTRACT
Diastereomerically pure alpha-trifluoromethyl alpha-amino nitriles obtained by Strecker-type reactions from chiral CF3 imines and iminium proved to be very attractive versatile intermediates for the synthesis of various alpha-trifluoromethyl amino compounds. From these synthons, both enantiomers of alpha-trifluoromethyl alanine, trifluoromethyl 1,2-diamines, and amino alcohols were conveniently obtained in enantiopure form in high yields in a few steps.
Sigma-Aldrich
1,1,1-Trifluoroacetone, 97%