Merck
  • Synthesis of 4-aryl-2-pyrrolidones and beta-aryl-gamma-amino-butyric acid (GABA) analogues by Heck arylation of 3-pyrrolines with arenediazonium tetrafluoroborates. Synthesis of (+/-)-rolipram on a multigram scale and chromatographic resolution by semipreparative chiral simulated moving bed chromatography.
Synthesis of 4-aryl-2-pyrrolidones and beta-aryl-gamma-amino-butyric acid (GABA) analogues by Heck arylation of 3-pyrrolines with arenediazonium tetrafluoroborates. Synthesis of (+/-)-rolipram on a multigram scale and chromatographic resolution by semipreparative chiral simulated moving bed chromatography.
The Journal of organic chemistry (2005-01-29)
Ariel L L Garcia, Marcos J S Carpes, Antonio C B M de Oca, Marco A G dos Santos, César C Santana, Carlos Roque D Correia
ABSTRACT
We report herein a new, practical, and economic synthesis of the phosphodiesterase inhibitor Rolipram on a multigram scale as well as the synthesis of new 4-aryl pyrrolidones and beta-aryl-gamma-amino butyric acids (GABA derivatives) employing an efficient Heck-Matsuda arylation of 3-pyrroline with aryldiazonium tetrafluoroborates. Racemic Rolipram was resolved into its enantiomers using chiral simulated moving bed chromatography having the low-cost microcrystalline cellulose triacetate as a chiral stationary phase.
Sigma-Aldrich
3,4-Dehydro-DL-proline, 98%
Sigma-Aldrich
3,4-Dehydro-L-proline
Sigma-Aldrich
N-Boc-2,5-dihydro-1H-pyrrole, 96%