Merck
Genetic and functional study of the IPO5 gene in schizophrenia.
Psychiatry research (2010-06-15)
Zhen-Qi Wang, Yang Liu, Ning Wu, Qi Xu, Shun-Zi Jin, Gui-Zhi Ju, Lin Ye, Li-Bo Liu, Xuan Zhang, Jiang Wu
ABSTRACT
The present work reported on a weak association of the importin 5 (IPO5) gene with schizophrenia in combined family and case-control samples and also investigated a possible mechanism by which the IPO5 gene may contribute to the development of the disease in a Chinese population. Our results suggest that abnormal expression and alternative splicing of the IPO5 gene may be involved in the pathophysiology of schizophrenia.