Merck
  • Expression of cytochrome P450 2A13 in human non-small cell lung cancer and its clinical significance.
Expression of cytochrome P450 2A13 in human non-small cell lung cancer and its clinical significance.
Journal of biomedical research (2013-05-31)
Li Sun, Xiaoli Fan
ABSTRACT
Lung cancer is one of the most important causes of cancer-related mortality worldwide. Human cytochrome P450 2A13 enzyme (CYP2A13) is predominantly expressed in the respiratory tract and could catalyze various carcinogens. In this study, we quantified CYP2A13 expression in non-small cell lung cancer (NSCLC) tissues and examined the relation between CYP2A13 and clinicopathologic factors. Thirty-five paired lung cancer and normal tissues were studied for the expression of the CYP2A13 gene by using real-time PCR and Western blotting assays. We also investigated the relationship between CYP2A13 expression and clinicopathologic factors such as age, gender, histology and lymph node status in tumor tissues. SPSS (17.0) statistical software was applied for data analysis. The real-time PCR results showed that there was no significant difference in the CYP2A13 mRNA transcript levels between tumor and paired normal tissues in the 35 samples and in 12 paired squamous cell carcinomas. In adenocarcinoma, the expression of CYP2A13 mRNA in tumor tissues was 12.5% of that in adjacent tissues (P < 0.05) and it was not associated with age, gender, histology and lymph node status of the patients. The amounts of CYP2A13 proteins detected by Western blotting assays correlated well with those of the corresponding mRNAs. In conclusion, the expression of CYP2A13 was downregulated in lung adenocarcinoma. CYP2A13 may be involved in the development and progression of lung adenocarcinoma.