Merck
Aβ 1-40 enhances the proliferation of human diploid fibroblasts.
Neurobiology of aging (2016-02-02)
Lindsey Theda, Michelle K Drews, Galynn Zitnik, Junko Oshima, George M Martin
ABSTRACT
There is a vast literature on the role of beta amyloid (Aβ) peptides in the pathogenesis of Alzheimer's disease. However, there is a paucity of research on the potential physiological functions of these evolutionarily conserved products of the Aβ precursor protein. Based on previous studies in neuroblastoma cells, we hypothesized that Aβ may contribute to the proliferation of somatic cells. We present evidence supporting this hypothesis for the case of cultured human skin fibroblasts immortalized with the catalytic subunit of human telomerase (hTERT). Optimal concentrations ranged from 100 pM-10 nM, depending on the nature of the assay.
Sigma-Aldrich
Amyloid β Protein Fragment 1-40, ≥90% (HPLC), powder