Merck
  • Glutathione S-transferase omega-2 polymorphism Asn142Asp modifies the risk of age-related cataract in smokers and subjects exposed to ultraviolet irradiation.
Glutathione S-transferase omega-2 polymorphism Asn142Asp modifies the risk of age-related cataract in smokers and subjects exposed to ultraviolet irradiation.
Clinical & experimental ophthalmology (2013-08-10)
Miroslav Stamenkovic, Tanja Radic, Ivan Stefanovic, Vesna Coric, Ivan Sencanic, Marija Pljesa-Ercegovac, Marija Matic, Vesna Jaksic, Tatjana Simic, Ana Savic-Radojevic
ABSTRACT
Glutathione S-transferase omega-1 and 2 have a unique range of enzymatic activities, including the regeneration of ascorbate by their dehydroascorbate reductase activities. Because these enzymes could have a protective role from oxidative damage in the lens, the question of whether the two coding glutathione S-transferase omega polymorphisms confer the risk of age-related cataract was addressed. rs4925 (Ala140Asp) of glutathione S-transferase omega-1 and rs156697 (Asn142Asp) of glutathione S-transferase omega-2 polymorphisms in 100 patients with age-related cataract and 130 controls were assessed. Presence of one mutant GSTO1*Asp or GSTO2*Asp allele did not contribute independently towards the risk of cataract; however, homozygous carriers of GSTO1*Asp/GSTO2*Asp haplotype demonstrated 3.42-fold enhanced risk of cataract development (95% confidence interval = 0.84-13.93; P = 0.086). When GSTO genotype was analysed in association with smoking or professional exposure to ultraviolet irradiation, carriers of at least one mutant GSTO2*Asp allele had increased risk of cataract development in comparison with individuals with wild-type GSTO2*Asn/Asn with no history of smoking or ultraviolet exposure (odds ratio = 6.89, 95% confidence interval = 1.81-16.21, P = 0.005; odds ratio = 4.10, 95% confidence interval = 1.23-13.74, P = 0.022, respectively). Regarding the distribution of particular glutathione S-transferase omega genotype and cataract type, the highest frequency of mutant GSTO2*Asp allele was found in patients with nuclear cataract. The results indicate that mutant GSTO2*Asp genotype is associated with increased risk of age-related cataract in smokers and ultraviolet-exposed subjects, suggesting a role of inefficient ascorbate regeneration in cataract development.