Merck
  • Polymerase I and transcript release factor acts as an essential modulator of glioblastoma chemoresistance.
Polymerase I and transcript release factor acts as an essential modulator of glioblastoma chemoresistance.
PloS one (2014-04-22)
Xin Wang, Tianzhu Liu, Yifeng Bai, Hongzhan Liao, Shengcong Qiu, Zhenhua Chang, Yanting Liu, Xiaohui Yan, Hongbo Guo
ABSTRACT
This study is to investigate if polymerase I and transcript release factor (PTRF) acts as a modulator in glioblastoma (GBM) chemoresistance. Multidrug resistant (MDR) GBM cell line U251AR was established by exposing the U251 cell line to imatinib. The 2D-DIGE and MALDI-TOF/TOF-MS were performed on U251 and U251AR cell lines to screen MDR-related proteins. The expression of PTRF was determined by Western blot and quantitative RT-PCR analyses. When compared with the parental U251 cells, expression of 21 proteins was significantly altered in U251AR cells. Among the 21 differentially expressed proteins, the expression of PTRF was up-regulated by 2.14 folds in U251AR cells when compared with that in the parental U251 cells. Knockdown of PTRF in GBM cell lines significantly increased chemosensitivity of cells to various chemical drugs and decreased the expression levels of caveolin1, a major structural component of caveolae. Expression levels of PTRF and caveolin1 were significantly up-regulated in the relapsed GBM patients. The mRNA level of PTRF and caveolin1 showed a positive correlation in the same GBM specimens. Our results indicate that PTRF acts as a modulator in GBM chemoresistance.