Merck
Oxidative alkoxycarbonylation of terminal alkenes with carbazates.
Chemical communications (Cambridge, England) (2013-06-14)
Yu-Han Su, Zhao Wu, Shi-Kai Tian
ABSTRACT
A range of terminal alkenes smoothly underwent palladium-catalyzed oxidative alkoxycarbonylation with carbazates under an oxygen atmosphere to afford structurally diverse α,β-unsaturated esters in moderate to good yields with excellent regioselectivity and E selectivity.
Sigma-Aldrich
Molecular sieves, 5 Å, beads, 8-12 mesh
Sigma-Aldrich
Molecular sieves, 5 Å, powder, undried