Merck
  • Two new gene cassettes, dfr17 (for trimethoprim resistance) and aadA4 (for spectinomycin/streptomycin resistance), inserted in an Escherichia coli class 1 integron.
Two new gene cassettes, dfr17 (for trimethoprim resistance) and aadA4 (for spectinomycin/streptomycin resistance), inserted in an Escherichia coli class 1 integron.
The Journal of antimicrobial chemotherapy (2000-07-07)
C Y Chang, L L Chang, Y H Chang, T M Lee, Y H Li, S F Chang
ABSTRACT
Two new gene cassettes, dfr17 and aadA4, inserted in a class 1 integron of Escherichia coli EC107, are described here. The dfr17 cassette encodes trimethoprim resistance and has 91% identity with the dfrVII dihydrofolate reductase gene. The aadA4 cassette confers resistance to spectinomycin and streptomycin and shows 94% identity with the aadA3 gene. The integron carrying the dfr17 and aadA4 cassettes was located on a conjugative plasmid, pEC1072.
Sigma-Aldrich
Spectinomycin, Ready Made Solution, 100 mg/mL in DMSO/H2O, 1:1