Merck
A Trigonal-Pyramidal Erbium(III) Single-Molecule Magnet.
Angewandte Chemie (International ed. in English) (2015-03-27)
Andrew J Brown, Dawid Pinkowicz, Mohamed R Saber, Kim R Dunbar
ABSTRACT
Given the recent advent of mononuclear single-molecule magnets (SMMs), a rational approach based on lanthanides with axially elongated f-electron charge cloud (prolate) has only recently received attention. We report herein a new SMM, [Li(THF)4[Er{N(SiMe3)2}3Cl]⋅2 THF, which exhibits slow relaxation of the magnetization under zero dc field with an effective barrier to the reversal of magnetization (ΔEeff/kB =63.3 K) and magnetic hysteresis up to 3 K at a magnetic field sweep rate of 34.6 Oe s(-1). This work questions the theory that oblate or prolate lanthanides must be stabilized with the appropriate ligand framework in order for SMM behavior to be favored.
Sigma-Aldrich
Eicosane, 99%