Merck
  • SP1-mediated microRNA-520d-5p suppresses tumor growth and metastasis in colorectal cancer by targeting CTHRC1.
SP1-mediated microRNA-520d-5p suppresses tumor growth and metastasis in colorectal cancer by targeting CTHRC1.
American journal of cancer research (2015-06-24)
Li Yan, Jiang Yu, Fei Tan, Geng-Tai Ye, Zhi-Yong Shen, Hao Liu, Yan Zhang, Jie-Fu Wang, Xian-Jun Zhu, Guo-Xin Li
ABSTRACT
Recent evidence suggests that miR-520 family has an important role in regulating tumorigenesis and development of various types of solid cancers. However, as one of the most common cancers in the world, there is little known about the underlying regulatory mechanisms of miR-520 in colorectal cancer (CRC). In the present study, we investigated the expression of microRNA-520d-5p (miR-520d-5p) in CRC specimens and then explored its potential role and mechanism in CRC progression. We found that miR-520d-5p was markedly down-regulated in CRC clinical specimens compared with adjacent normal tissues by real-time PCR. Dual-luciferase assays confirmed that miR-520d-5p directly targeting CTHRC1 and SP1 transactivate miR-520d-5p by binding to its upstream promoter region. The biological functional experiments showed that ectopic re-expression of miR-520d-5p suppressed CRC cell proliferation, migration and invasion, whereas the inhibition of miR-520d-5p displayed an inverse effect in vitro and in vivo. Western blot shown that miR-520d-5p abrogated the epithelial-mesenchymal transition by inactivating the phosphorylation of Erk1/2. In conclusion, our findings indicate that miR-520d-5p is significantly down-expressed and involved in CRC progression and metastasis by targeting CTHRC1 and regulated by SP1, which provide new support for miR-520d-5p maybe as a novel anti-onco molecular target for the treatment of CRC in the future.
Sigma-Aldrich
MISSION® esiRNA, targeting human SP1
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)