Merck
  • Impact of a functionalized olive oil extract on the uterus and the bone in a model of postmenopausal osteoporosis.
Impact of a functionalized olive oil extract on the uterus and the bone in a model of postmenopausal osteoporosis.
European journal of nutrition (2013-10-31)
Annekathrin Martina Keiler, Oliver Zierau, Ricardo Bernhardt, Dieter Scharnweber, Nikolaos Lemonakis, Aikaterini Termetzi, Leandros Skaltsounis, Günter Vollmer, Maria Halabalaki
ABSTRACT
The Mediterranean diet rich in fruits, vegetables and olive oil has been related to a lower osteoporosis incidence and accordingly to a reduced fracture risk. These observations might be mediated by the active constituents of extra virgin olive oil, and especially polyphenols. In the context of exploring the features of olive oil active constituents on postmenopausal osteoporosis, an extra virgin olive oil total polyphenolic fraction (TPF) was isolated and its effect on the bone loss attenuation was investigated. Female Lewis rats were ovariectomized and fed a diet enriched with a total phenolic extract of extra virgin olive oil in a concentration of 800 mg/kg diet. Oleocanthal, one compound of the polyphenolic fraction, showed a higher relative estrogen receptor binding affinity to the ERα compared to the ERβ. While the TPF only slightly induced the uterine wet weight (490.7 ± 53.7 vs. 432.7 ± 23, p = 0.058), TPF regulated estrogen response genes in the uterus (progesterone receptor, antigen identified by monoclonal antibody Ki67, complement C3). Comparing the quantified bone parameters, the oral TPF substitution did not attenuate the ovariectomy-induced bone loss. The administration of extra virgin olive oil polyphenols regulated uterine estrogen response marker genes in an E2-agonistic manner. The bone loss induced by estrogen ablation was not mitigated by treatment with the polyphenolic extract.
Sigma-Aldrich
Syringaldehyde, 98%
Sigma-Aldrich
Syringaldehyde, ≥98%, FG