Merck
  • Identification of a novel in-frame de novo mutation in SPTAN1 in intellectual disability and pontocerebellar atrophy.
Identification of a novel in-frame de novo mutation in SPTAN1 in intellectual disability and pontocerebellar atrophy.
European journal of human genetics : EJHG (2012-01-20)
Fadi F Hamdan, Hirotomo Saitsu, Kiyomi Nishiyama, Julie Gauthier, Sylvia Dobrzeniecka, Dan Spiegelman, Jean-Claude Lacaille, Jean-Claude Décarie, Naomichi Matsumoto, Guy A Rouleau, Jacques L Michaud
ABSTRACT
Heterozygous in-frame mutations (p.E2207del and p.R2308_M2309dup) in the α-II subunit of spectrin (SPTAN1) were recently identified in two patients with intellectual disability (ID), infantile spasms (IS), hypomyelination, and brain atrophy. These mutations affected the C-terminal domain of the protein, which contains the nucleation site of the α/β spectrin heterodimer. By screening SPTAN1 in 95 patients with idiopathic ID, we found a de novo in-frame mutation (p.Q2202del) in the same C-terminal domain in a patient with mild generalized epilepsy and pontocerebellar atrophy, but without IS, hypomyelination, or other brain structural defects, allowing us to define the core phenotype associated with these C-terminal SPTAN1 mutations. We also found a de novo missense variant (p.R566P) of unclear clinical significance in a patient with non-syndromic ID. These two mutations induced different patterns of aggregation between spectrin subunits in transfected neuronal cell lines, providing a paradigm for the classification of candidate variants.