Merck
  • Regioselective facile one-pot Friedländer synthesis of sugar-based heterocyclic biomolecules.
Regioselective facile one-pot Friedländer synthesis of sugar-based heterocyclic biomolecules.
Carbohydrate research (2010-08-07)
Subbiah Nagarajan, Pandian Arjun, Nanjian Raaman, Thangamuthu Mohan Das
ABSTRACT
Regioselective facile one-pot synthesis of 16 different sugar-based quinoline, naphthyridine, and xanthone derivatives is reported. The compounds are characterized by NMR spectroscopy and elemental analysis. The beta-Anomeric form of the sugar moiety was identified from (1)H NMR studies. Antimicrobial studies of these sugar-heterocyclic derivatives, 3a, 3b, 3f, 5c, 7a, 7b, and 7c show excellent activity against different microbes.
Sigma-Aldrich
2-Amino-3,5-dibromobenzaldehyde, 97%