- (Photo)physical Properties of New Molecular Glasses End-Capped with Thiophene Rings Composed of Diimide and Imine Units.
(Photo)physical Properties of New Molecular Glasses End-Capped with Thiophene Rings Composed of Diimide and Imine Units.
The journal of physical chemistry. C, Nanomaterials and interfaces (2014-06-27)
Marzena Grucela-Zajac, Katarzyna Bijak, Slawomir Kula, Michal Filapek, Malgorzata Wiacek, Henryk Janeczek, Lukasz Skorka, Jacek Gasiorowski, Kurt Hingerl, Niyazi Serdar Sariciftci, Natalia Nosidlak, Gabriela Lewinska, Jerzy Sanetra, Ewa Schab-Balcerzak
PMID24966893
ABSTRACT
New symmetrical arylene bisimide derivatives formed by using electron-donating-electron-accepting systems were synthesized. They consist of a phthalic diimide or naphthalenediimide core and imine linkages and are end-capped with thiophene, bithiophene, and (ethylenedioxy)thiophene units. Moreover, polymers were obtained from a new diamine,