Merck
  • Fine analysis of spontaneous MAGE-C1/CT7-specific immunity in melanoma patients.
Fine analysis of spontaneous MAGE-C1/CT7-specific immunity in melanoma patients.
Proceedings of the National Academy of Sciences of the United States of America (2010-08-11)
Natko Nuber, Alessandra Curioni-Fontecedro, Claudia Matter, Davide Soldini, Jean Marie Tiercy, Lotta von Boehmer, Holger Moch, Reinhard Dummer, Alexander Knuth, Maries van den Broek
ABSTRACT
Cancer/testis (CT) antigens represent prime candidates for immunotherapy in cancer patients, because their expression is restricted to cancer cells and germ cells of the testis. MAGE-C1/CT7 is a CT antigen that is highly expressed in several types of cancers. Spontaneous occurrence of CT7-specific antibodies was previously detected by SEREX screen in a melanoma patient. However, naturally occurring CT7-specific T-cell responses have thus far not been detected. Peripheral blood mononuclear cells (PBMCs) from 26 metastatic melanoma patients expressing CT7 in their tumor lesions (CT7(+)) were analyzed for CT7-specific T-cell responses using overlapping peptides. CT7-specific CD4(+) T-cell responses were detected in three patients (11.5%). These CT7-specific CD4(+) T-cell responses were detectable in melanoma patients' PBMCs exclusively from preexisting CD45RA(-) memory CD4(+) T-cell pool. Additional CT7-specific memory CD4(+) T-cell responses were detected in CT7(+) melanoma patients after depletion of CD4(+)CD25high Treg cells showing that Treg cells impact on CT7-specific CD4(+) T cells in melanoma patients. CT7-specific CD4(+) T-cell clones were generated and used to define minimal epitopes, restriction elements, and confirm the recognition of naturally processed antigen. Surprisingly, these clones were able to secrete perforin and exert cytotoxicity. This study shows that CT7 can induce specific cellular immunity in melanoma patients. Based on these findings, CT7 will be further explored as a potential vaccine for melanoma immunotherapy.