Merck
  • A practical synthesis of long-chain iso-fatty acids (iso-C12-C19) and related natural products.
A practical synthesis of long-chain iso-fatty acids (iso-C12-C19) and related natural products.
Beilstein journal of organic chemistry (2013-09-26)
Mark B Richardson, Spencer J Williams
ABSTRACT
A gram-scale synthesis of terminally-branched iso-fatty acids (iso-C12-C19) was developed commencing with methyl undec-10-enoate (methyl undecylenate) (for iso-C12-C14) or the C15 and C16 lactones pentadecanolide (for iso-C15-C17) and hexadecanolide (for iso-C18-C19). Central to the approaches outlined is the two-step construction of the terminal isopropyl group through addition of methylmagnesium bromide to the ester/lactones and selective reduction of the resulting tertiary alcohols. Thus, the C12, C17 and C18 iso-fatty acids were obtained in three steps from commercially-available starting materials, and the remaining C13-C16 and C19 iso-fatty acids were prepared by homologation or recursive dehomologations of these fatty acids or through intercepting appropriate intermediates. Highlighting the synthetic potential of the iso-fatty acids and various intermediates prepared herein, we describe the synthesis of the natural products (S)-2,15-dimethylpalmitic acid, (S)-2-hydroxy-15-methylpalmitic acid, and 2-oxo-14-methylpentadecane.
Sigma-Aldrich
Pentadecanolide, 98%