- ZDHHC8 gene may play a role in cortical volumes of patients with schizophrenia.
ZDHHC8 gene may play a role in cortical volumes of patients with schizophrenia.
Schizophrenia research (2013-02-14)
Vanessa K Ota, Ary Gadelha, Idaiane B Assunção, Marcos L Santoro, Denise M Christofolini, Fernanda T Bellucco, Airton F Santos-Filho, Gustavo L Ottoni, Diogo R Lara, Jair J Mari, Maria I Melaragno, Marília A C Smith, Rodrigo A Bressan, Sintia I Belangero, Andrea P Jackowski
PMID23403413
ABSTRACT
ZDHHC8 rs175174 polymorphism is located in 22q11.2 region and its role in brain volume has not been fully addressed. A total of 282 schizophrenia patients and 379 controls were genotyped. A sample of 138 patients underwent brain MRI scan. No association was found between schizophrenia and genotypes. Nevertheless, GG-genotype carriers presented gray matter volume (GMV) reduction in frontal lobe compared to A-allele carriers, and cerebellar hemispheres GMV reductions were found in G-allele carriers compared to AA-genotype. Moreover, A-allele carriers presented posterior brain GMV reductions when compared to GG-genotype. These data suggest that ZDHHC8 may play a role in cortical volumes.