Merck
  • Bisphenol A in combination with insulin can accelerate the conversion of 3T3-L1 fibroblasts to adipocytes.
Bisphenol A in combination with insulin can accelerate the conversion of 3T3-L1 fibroblasts to adipocytes.
Journal of lipid research (2002-04-25)
Hiroshi Masuno, Teruki Kidani, Keizo Sekiya, Kenshi Sakayama, Takahiko Shiosaka, Haruyasu Yamamoto, Katsuhisa Honda
ABSTRACT
The confluent cultures of 3T3-L1 fibroblasts were treated with or without bisphenol A (BPA) for 2 days and subsequently treated with insulin (INS) alone for 9 days. When BPA was absent during the first 2-day treatment period, the cultures contained 1.6 microg/microg DNA of triacylglycerol (TG), 202 mU/mg DNA of lipoprotein lipase (LPL) activity, and 462 nmol/min/mg DNA of glycerol-3-phosphate dehydrogenase (GPDH) activity. The presence of BPA during the same period caused a 150% increase in the TG content, a 60% increase in the LPL activity, and a 500% increase in the GPDH activity. Thus, BPA by itself can trigger 3T3-L1 fibroblasts to differentiate into adipocytes. Next, the confluent cultures were treated with BPA for 2 days and subsequently treated with a combination of INS and BPA for 9 days. The simultaneous presence of BPA with INS caused a 370% increase in the TG content, a 200% increase in the LPL activity, and a 225% increase in the GPDH activity compared with the cultures treated with INS alone. The amount of [(3)H]thymidine incorporated into DNA was lower in the cultures treated with INS in the presence of BPA than in those treated with INS alone, indicating that BPA has an anti-proliferative activity on 3T3-L1 cells. Taken together, our results indicate that BPA in combination with INS can accelerate the adipocyte conversion.
Sigma-Aldrich
Bisphenol A bis(chloroformate), 95%