- Doublecortin, a brain-specific gene mutated in human X-linked lissencephaly and double cortex syndrome, encodes a putative signaling protein.
Doublecortin, a brain-specific gene mutated in human X-linked lissencephaly and double cortex syndrome, encodes a putative signaling protein.
X-linked lissencephaly and "double cortex" are allelic human disorders mapping to Xq22.3-Xq23 associated with arrest of migrating cerebral cortical neurons. We identified a novel 10 kb brain-specific cDNA interrupted by a balanced translocation in an XLIS patient that encodes a novel 40 kDa predicted protein named Doublecortin. Four double cortex/X-linked lissencephaly families and three sporadic double cortex patients show independent doublecortin mutations, at least one of them a de novo mutation. Doublecortin contains a consensus Abl phosphorylation site and other sites of potential phosphorylation. Although Doublecortin does not contain a kinase domain, it is homologous to the amino terminus of a predicted kinase protein, indicating a likely role in signal transduction. Doublecortin, along with the newly characterized mDab1, may define an Abl-dependent pathway regulating neuronal migration.