Merck
The DEAD-box RNA helicase Dbp5 functions in translation termination.
Science (New York, N.Y.) (2007-02-03)
Thomas Gross, Anja Siepmann, Dorotheé Sturm, Merle Windgassen, John J Scarcelli, Matthias Seedorf, Charles N Cole, Heike Krebber
ABSTRACT
In eukaryotes, termination of messenger RNA (mRNA) translation is mediated by the release factors eRF1 and eRF3. Using Saccharomyces cerevisiae as a model organism, we have identified a member of the DEAD-box protein (DBP) family, the DEAD-box RNA helicase and mRNA export factor Dbp5, as a player in translation termination. Dbp5 interacts genetically with both release factors and the polyadenlyate-binding protein Pab1. A physical interaction was specifically detected with eRF1. Moreover, we show that the helicase activity of Dbp5 is required for efficient stop-codon recognition, and intact Dbp5 is essential for recruitment of eRF3 into termination complexes. Therefore, Dbp5 controls the eRF3-eRF1 interaction and thus eRF3-mediated downstream events.