- Sequential continuous flow processes for the oxidation of amines and azides by using HOF·MeCN.
Sequential continuous flow processes for the oxidation of amines and azides by using HOF·MeCN.
ChemSusChem (2011-12-16)
Christopher B McPake, Christopher B Murray, Graham Sandford
PMID22170755
ABSTRACT
The generation and use of the highly potent oxidising agent HOF·MeCN in a controlled single continuous flow process is described. Oxidations of amines and azides to corresponding nitrated systems by using fluorine gas, water and acetonitrile by sequential gas-liquid/liquid-liquid continuous flow procedures are reported.