Merck
  • Towards dynamic control of wettability by using functionalized altitudinal molecular motors on solid surfaces.
Towards dynamic control of wettability by using functionalized altitudinal molecular motors on solid surfaces.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-06-21)
Gábor London, Kuang-Yen Chen, Gregory T Carroll, Ben L Feringa
ABSTRACT
We report the synthesis of altitudinal molecular motors that contain functional groups in their rotor part. In an approach to achieve dynamic control over the properties of solid surfaces, a hydrophobic perfluorobutyl chain and a relatively hydrophilic cyano group were introduced to the rotor part of the motors. Molecular motors were attached to quartz surfaces by using interfacial 1,3-dipolar cycloadditions. To test the effect of the functional groups on the rotary motion, photochemical and thermal isomerization studies of the motors were performed both in solution and when attached to the surface. We found that the substituents have no significant effect on the thermal and photochemical processes, and the functionalized motors preserved their rotary function both in solution and on a quartz surface. Preliminary results on the influence of the functional groups on surface wettability are also described.