Merck
  • Evaluation of chelating ion-exchange resins for separating Cr(III) from industrial effluents.
Evaluation of chelating ion-exchange resins for separating Cr(III) from industrial effluents.
Journal of hazardous materials (2009-05-02)
Sofia A Cavaco, Sandra Fernandes, Cátia M Augusto, Margarida J Quina, Licínio M Gando-Ferreira
ABSTRACT
In this study two chelating resins containing iminodiacetic acid groups (Amberlite IRC 748 and Diaion CR 11) and a chelating resin based on sulfonic and diphosphonic acid groups (Diphonix) were investigated in order to separate Cr(III) from industrial effluents produced in hard and decorative electroplating. Samples of two industrial plants were characterized during a period of about one year and a half in terms of the metals content (Cr, Cu, Na, Ca, Fe and Ni), Total Suspended Solids (TSS), Total Dissolved Solids (TDS), Chemical Oxygen Demand (COD) and pH. Some of the physical properties of the resins, namely the moisture content, apparent density, intraparticle porosity and the particle size distribution were also evaluated. To quantify the sorption capacity of the resins, batch experiments were performed using synthetic solutions of Cr(III), as well as solutions of Fe in the case of Diphonix. The Langmuir and Langmuir-Freundlich isotherms enabled a good description of the ion-exchange equilibrium data, and the maximum sorption capacity determined for Amberlite and Diaion was 3.6 mequiv./g(dry resin). For Diphonix that parameter was 3.4 mequiv./g(dry resin). The Diphonix resin exhibits a high selectivity for transition metals (Fe, Ni) over the chromium trivalent. Therefore, it was screened as the most suitable for selectively removing those metal impurities from chromium electroplating effluents. For this resin, the sorption capacity is strongly dependent on the initial pH of the solution. Though, high regeneration efficiencies of Diphonix for stripping Cr(III) were found by using a mixture of NaOH/H(2)O(2). The mathematical model tested for describing the dynamics of the process allowed a good fitting to the experimental data and enabled the estimation of effective pore diffusivity of Cr(III). The saturations of Diphonix with industrial effluents demonstrated that the breakthrough capacity of the resin is affected by the presence of other species in solution, such as Fe and Ni. Nevertheless, these effluents may be treated with this resin, being possible to separate Cr(III) from other transition metallic ions in solution.
Product Description
Supelco
Diaion® CR11 sodium form, pkg of 1000 g