Merck
  • Quantitative proteomics analysis highlights the role of redox hemostasis and energy metabolism in human embryonic stem cell differentiation to neural cells.
Quantitative proteomics analysis highlights the role of redox hemostasis and energy metabolism in human embryonic stem cell differentiation to neural cells.
Journal of proteomics (2014-02-18)
Ali Fathi, Maryam Hatami, Haghighat Vakilian, Chia-Li Han, Yu-Ju Chen, Hossein Baharvand, Ghasem Hosseini Salekdeh
ABSTRACT
Neural differentiation of human embryonic stem cells (hESCs) is a unique opportunity for in vitro analyses of neurogenesis in humans. Extrinsic cues through neural plate formation are well described in the hESCs although intracellular mechanisms underlying neural development are largely unknown. Proteome analysis of hESC differentiation to neural cells will help to further define molecular mechanisms involved in neurogenesis in humans. Using a two-dimensional differential gel electrophoresis (2D-DIGE) system, we analyzed the proteome of hESC differentiation to neurons at three stages, early neural differentiation, neural ectoderm and mature neurons. Out of 137 differentially accumulated protein spots, 118 spots were identified using MALDI-TOF/TOF and LC MS/MS. We observed that proteins involved in redox hemostasis, vitamin and energy metabolism and ubiquitin dependent proteolysis were more abundant in differentiated cells, whereas the abundance of proteins associated with RNA processing and protein folding was higher in hESCs. Higher abundance of proteins involved in maintaining cellular redox state suggests the importance of redox hemostasis in neural differentiation. Furthermore, our results support the concept of a coupling mechanism between neuronal activity and glucose utilization. The protein network analysis showed that the majority of the interacting proteins were associated with the cell cycle and cellular proliferation. These results enhanced our understanding of the molecular dynamics that underlie neural commitment and differentiation. In highlighting the role of redox and unique metabolic properties of neuronal cells, the present findings add insight to our understanding of hESC differentiation to neurons. The abundance of fourteen proteins involved in maintaining cellular redox state, including 10 members of peroxiredoxin (Prdx) family, mainly increased during differentiation, thus highlighting a link of neural differentiation to redox. Our results revealed markedly higher expression of genes encoding enzymes involved in the glycolysis and amino acid synthesis during differentiation. Protein network analysis predicted a number of critical mediators in hESC differentiation. These proteins included TP53, CTNNB1, SMARCA4, TNF, TERT, E2F1, MYC, RB1, and AR.
Sigma-Aldrich
Western Blocker Solution, for HRP detection systems
Pricing and availability is not currently available.
Sigma-Aldrich
Fibronectin rat plasma, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.