Merck
  • Two ellagitannins from the leaves of Terminalia triflora with inhibitory activity on HIV-1 reverse transcriptase.
Two ellagitannins from the leaves of Terminalia triflora with inhibitory activity on HIV-1 reverse transcriptase.
Phytotherapy research : PTR (2004-10-09)
V Martino, J Morales, J J Martínez-Irujo, M Font, A Monge, J Coussio
ABSTRACT
The bioassay- guided fractionation of the aqueous extract of Terminalia triflora leaves afforded punicalin and 2-O-galloylpunicalin, isolated for the first time from this species. These compounds showed inhibitory activity on HIV-1 reverse transcriptase in a dose-dependent manner. Punicalin showed an IC(50) of 0.11 microg/ml (0.14 microM) and 2-O-galloylpunicalin an IC(50) of 0.10 microg/ml (0.11 microM).