Merck
  • Synthesis and antiviral activity of new pyrazole and thiazole derivatives.
Synthesis and antiviral activity of new pyrazole and thiazole derivatives.
European journal of medicinal chemistry (2009-05-08)
Osama I el-Sabbagh, Mohamed M Baraka, Samy M Ibrahim, Christophe Pannecouque, Graciela Andrei, Robert Snoeck, Jan Balzarini, Adel A Rashad
ABSTRACT
New N-acetyl (5-8) and N-thiocarbamoyl (9-12) derivatives of 4,5-dihydropyrazole were synthesized starting from alpha,beta-unsaturated ketones under the effect of hydrazine hydrate and thiosemicarbazide, respectively. N-Thiocarbamoylpyrazole derivatives (9-12) were cyclized using either ethyl bromoacetate or phenacyl bromides to afford the novel pyrazolothiazol-4(5H)-ones (13-16) or pyrazolothiazoles (17-24). The antiviral activity for such novel compounds against a broad panel of viruses in different cell cultures revealed that N-acetyl 4,5-dihydropyrazole 7 was the only active one at subtoxic concentrations against vaccinia virus (Lederle strain) in HEL cell cultures with a 50% effective concentration (EC(50)) value of 7 microg/ml.
Sigma-Aldrich
Ethyl bromoacetate, purum, ≥97.0% (GC)