Merck
  • Heterocyclic methacrylates for clinical applications. II. Room temperature polymerizing systems for potential clinical use.
Heterocyclic methacrylates for clinical applications. II. Room temperature polymerizing systems for potential clinical use.
Biomaterials (1991-09-01)
M P Patel, M Braden
ABSTRACT
A number of useful room temperature polymerizing resins were formulated, based on poly(ethyl methacrylate) powder and a range of low shrinkage heterocyclic methacrylate monomers. N,N-dimethyl-p-toluidine or p-tolyl diethanolamine were used as activating amines, but the latter material is less active and some care is necessary with respect to the source of the monomer. Isobornyl methacrylate is a useful diluent monomer to reduce the exotherm.
Sigma-Aldrich
2,2′-(4-Methylphenylimino)diethanol, ≥97.0% (NT)