Merck
  • Effects of select nitrocompounds on in vitro ruminal fermentation during conditions of limiting or excess added reductant.
Effects of select nitrocompounds on in vitro ruminal fermentation during conditions of limiting or excess added reductant.
Bioresource technology (2008-06-10)
Robin C Anderson, Nathan A Krueger, Thaddeus B Stanton, Todd R Callaway, Thomas S Edrington, Roger B Harvey, Yong Soo Jung, David J Nisbet
ABSTRACT
Ruminal methane (CH(4)) production results in the loss of up to 12% of gross energy intake and contributes nearly 20% of the United States' annual emission of this greenhouse gas. We report the effects of select nitrocompounds on ruminal fermentation after 22 h in vitro incubation (39 degrees C) with or without additions of hydrogen (H(2)), formate or both. In incubations containing no added reductant, CH(4) production was inhibited 41% by 2-nitro-1-propanol (2NPOH) and >97% by 3-nitro-1-propionic acid (3NPA), nitroethane (NE) and 2-nitroethanol (2NEOH) compared to non-treated controls and H(2) did not accumulate. With formate as the sole added reductant, nitro-treatment reduced CH(4) production by >99% and caused 42% to complete inhibition of formate catabolism compared to controls, and the accumulation of H(2) increased slightly. Nitro-treatment decreased CH(4) production 57-98% from that of controls when supplied H(2) or formate plus H(2). Formate catabolism was decreased 42-84% from that in controls by all nitro-treatments except 3NPA with both formate and H(2). Greater than 97% of the added H(2) was catabolized within controls; >84% was catabolized in nitro-treated incubations. Acetate, propionate and butyrate accumulations were unaffected by nitro-treatment irregardless of reductant; however, effects on ammonia and branched chain fatty acid accumulations varied. These results suggest that nitro-treatment inhibited formate dehydrogenase/formate hydrogen lyase and hydrogenase activity.
Sigma-Aldrich
2-Nitroethanol, 97%