Merck
  • A cell-free protein synthesis system as an investigational tool for the translation stop processes.
A cell-free protein synthesis system as an investigational tool for the translation stop processes.
FEBS letters (2002-06-14)
Taek Jin Kang, Ji Hyoung Woo, Hui Kyoung Song, Jin Ho Ahn, Jae Wook Kum, Jin Han, Cha Yong Choi, Hyun Joo
ABSTRACT
Using Escherichia coli cell-free protein synthesis system and aminoacylated amber suppressor tRNA, we successfully inserted an unnatural amino acid S-(2-nitrobenzyl)cysteine into human erythropoietin. Three different types of translation stop suppression were observed and each of the three types was easily discerned with SDS-PAGE. Optimal conditions were established for correct stop and programmed suppressions. Since this system differentiates proteins produced by misreading of codons from those produced by programmed suppression, we conclude that this cell-free translation system that we describe in this paper will be of a great use for future investigations on translation stop processes.