Merck
Disposition and metabolic fate of 14C-quazepam in man.
Drug metabolism and disposition: the biological fate of chemicals (1985-01-01)
N Zampaglione, J M Hilbert, J Ning, M Chung, R Gural, S Symchowicz
ABSTRACT
The absorption, metabolism, and excretion of quazepam, a new benzodiazepine hypnotic, was investigated in six normal male volunteers after oral administration of 25 mg 14C-quazepam in solution. Quazepam was well absorbed. Plasma radioactivity peaked (324.6 ng quazepam eq/ml) 1.75 hr postdose. Unchanged quazepam reached its maximum plasma level (148 ng/ml) at 1.5 hr with an apparent absorption half-life of 0.4 hr. Major plasma metabolites of quazepam were 2-oxoquazepam (OQ), obtained by replacement of S by O,N-desalkyl-2-oxoquazepam (DOQ), and 3-hydroxy-2-oxoquazepam (HOQ) glucuronide. Both OQ and DOQ are pharmacologically active. Plasma elimination half-lives for quazepam, OQ, DOQ, and radioactivity were 39, 40, 69, and 76 hr, respectively. The respective AUC (120 hr) values were 715, 438, 3323, and 11402 hr X ng/ml. Approximately 54% of the radioactive dose was excreted in the urine (31.3%) and feces (22.7%) over a 5-day period. HOQ glucuronide was the major urinary metabolite of quazepam. Other metabolites present in the urine in relatively large amounts were glucuronides of DOQ and HDOQ.
Product Description
Supelco
Desalkylflurazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®