Merck
  • Asymmetric synthesis of trifluoromethylated amines via catalytic enantioselective isomerization of imines.
Asymmetric synthesis of trifluoromethylated amines via catalytic enantioselective isomerization of imines.
Journal of the American Chemical Society (2012-08-22)
Yongwei Wu, Li Deng
ABSTRACT
A new approach toward the asymmetric synthesis of optically active trifluoromethylated amines was enabled by an unprecedented, highly enantioselective catalytic isomerization of trifluoromethyl imines with a new chiral organic catalyst. Not only aryl but also alkyl trifluoromethylated amines could be obtained in high enantioselectivities.