Merck
  • Complex phase separation in poly(acrylonitrile-butadiene-styrene)-modified epoxy/4,4'-diaminodiphenyl sulfone blends: generation of new micro- and nanosubstructures.
Complex phase separation in poly(acrylonitrile-butadiene-styrene)-modified epoxy/4,4'-diaminodiphenyl sulfone blends: generation of new micro- and nanosubstructures.
The journal of physical chemistry. B (2009-04-01)
P Jyotishkumar, Joachim Koetz, Brigitte Tiersch, Veronika Strehmel, Ceren Ozdilek, Paula Moldenaers, Rudiger Hässler, Sabu Thomas
ABSTRACT
The epoxy system containing diglycidyl ether of bisphenol A and 4,4'-diaminodiphenyl sulfone is modified with poly(acrylonitrile-butadiene-styrene) (ABS) to explore the effects of the ABS content on the phase morphology, mechanism of phase separation, and viscoelastic properties. The amount of ABS in the blends was 5, 10, 15, and 20 parts per hundred of epoxy resin (phr). Transmission electron microscopy (TEM) and scanning electron microscopy (SEM) were employed to investigate the final morphology of ABS-modified epoxy blends. Scanning electron microscopic studies of 15 phr ABS-modified epoxy blends reveal a bicontinuous structure in which both epoxy and ABS are continuous, with substructures of the ABS phase dispersed in the continuous epoxy phase and substructures of the epoxy phase dispersed in the continuous ABS phase. TEM micrographs of 15 phr ABS-modified epoxy blends confirm the results observed by SEM. TEM micrographs reveal the existence of nanosubstructures of ABS in 20 phr ABS-modified epoxy blends. To the best of our knowledge, to date, nanosubstructures have never been reported in any epoxy/thermoplastic blends. The influence of the concentration of the thermoplastic on the generated morphology as analyzed by SEM and TEM was explained in detail. The evolution and mechanism of phase separation was investigated in detail by optical microscopy (OM) and small-angle laser light scattering (SALLS). At concentrations lower than 10 phr the system phase separates through nucleation and growth (NG). However, at higher concentrations, 15 and 20 phr, the blends phase separate through both NG and spinodal decomposition mechanisms. On the basis of OM and SALLS, we conclude that the phenomenon of complex substructure formation in dynamic asymmetric blends is due to the combined effect of hydrodynamics and viscoelasticity. Additionally, dynamic mechanical analysis was carried out to evaluate the viscoelastic behavior of the cross-linked epoxy/ABS blends. Finally, apparent weight fractions of epoxy and ABS components in epoxy- and ABS-rich phases were evaluated from T(g) analysis.