Merck
  • [Effects of diclofenac sodium combined with dionine in cases with fibrinous membrane after intraocular lens implantation].
[Effects of diclofenac sodium combined with dionine in cases with fibrinous membrane after intraocular lens implantation].
Hunan yi ke da xue xue bao = Hunan yike daxue xuebao = Bulletin of Hunan Medical University (2003-01-23)
S B Jia, L S Tang
ABSTRACT
The effects of diclofenac sodium(DFNa) combined with dionine in cases with fibrin exudation membrane on intraocular lens (IOL) were studied. Thirty-two eyes, derived from sixteen adult pure bred New Zealand rabbits, were divided at random into two groups after extracapsular lens extraction with posterior chamber IOL implantation: (1) rabbits received DFNa eyedrops combined with dionine eyedrops; (2) rabbits received Pred forte eyedrops. The sum of fibrinous exudation membrane on IOL was observed on 1, 3, 5, 7, 14, 21, 30d postoperation. The fibrin exudations in the DFNa combined with dionine group is less than the pred forte group (P > 0.05). DFNa combined with dionine is effective in treating fibrin exudation membrane after extracapsular lens extraction and IOL implantation, and it is more effective than the pred forte.
Product Description
Supelco
Ethylmorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®