Skip to Content
Beginning October 1st, 2024, the Life Science business of Merck Ltd. Taiwan will implement list price increases on our Science and Lab Solutions products. Learn More
Merck
  • [Moroxidine, an antiviral agent used for the treatment of shingles (herpes zoster)].
[Moroxidine, an antiviral agent used for the treatment of shingles (herpes zoster)].
Romanian journal of virology (1993-01-01)
P Athanasiu, A Petrescu, V Vulcan
ABSTRACT
Treatment with moroxidine, Romanian preparation with virustatic effects, was applied in 350 patients with different localisation herpes zoster lesions. Treatment had good effects, especially when it was applied early.
Sigma-Aldrich
Moroxydine hydrochloride, 99%
Pricing and availability is not currently available.