Merck
  • Dephosphorylation and benzoylation reactions occurring in the benzoolysis of diacyl-, alkylacyl-, alk-1-enylacyl-, and acyl-lyso-glycerophosphocholines.
Dephosphorylation and benzoylation reactions occurring in the benzoolysis of diacyl-, alkylacyl-, alk-1-enylacyl-, and acyl-lyso-glycerophosphocholines.
Journal of lipid research (1997-08-01)
W J Gelsema, H van den Bosch
ABSTRACT
Series of benzoolysis experiments with 1,2-dipalmitoyl glycerophosphocholine (GPC), 1-palmitoyl-2-lyso GPC, and 1-O-hexadecenyl-2-oleoyl GPC, as well as a few experiments with 1-oleoyl-2-acetyl GPC and 1-O-hexadecyl-2-acetyl GPC are reported. The results are used to present a consistent picture of the various dephosphorylation and benzoylation reactions occurring in molten benzoic anhydride.
Sigma-Aldrich
Benzoic anhydride, ≥95%