Merck
  • Novel inhibitors of prolyl 4-hydroxylase. 5. The intriguing structure-activity relationships seen with 2,2'-bipyridine and its 5,5'-dicarboxylic acid derivatives.
Novel inhibitors of prolyl 4-hydroxylase. 5. The intriguing structure-activity relationships seen with 2,2'-bipyridine and its 5,5'-dicarboxylic acid derivatives.
Journal of medicinal chemistry (1993-11-26)
N J Hales, J F Beattie
ABSTRACT
Members of a series of 2,2'-bipyridines have been synthesized and tested as inhibitors of prolyl hydroxylase (EC 1.14.11.2). The structure-activity relationships seen with [2,2'-bipyridine]-5-carboxylic acid (4) closely resemble those of pyridine-2-carboxylic acid (2). Accordingly, [2,2'-bipyridine]-5,5'-dicarboxylic acid (11, IC50 = 0.19 microM) is the most potent inhibitor of its type yet reported. However, 2,2'-bipyridines lacking a 5-carboxylate are poor inhibitors. These contrasting structure-activity relationships are discussed in terms of net anionic charge, iron chelation, and the availability of alternative putative binding modes at a single binding site in each catalytic subunit.
Sigma-Aldrich
2,2′-Bipyridine-5,5′-dicarboxylic acid, 97%