Merck
  • Risk factors for a low linezolid trough plasma concentration in acute infections.
Risk factors for a low linezolid trough plasma concentration in acute infections.
Antimicrobial agents and chemotherapy (2013-02-14)
Laura Morata, Marta Cuesta, Jhon F Rojas, Sebastian Rodriguez, Merce Brunet, Gregori Casals, Nazareth Cobos, Cristina Hernandez, José A Martínez, Josep Mensa, Alex Soriano
ABSTRACT
Linezolid is an antibiotic with time-dependent activity, and both the percentage of time that plasma concentrations exceed the MIC and the area under the concentration-time curve over 24 h in the steady state divided by the MIC (AUC24/MIC ratio) are associated with clinical response. The aim of this study was to analyze the linezolid trough plasma concentration (C(min)) and to determine factors associated with a C(min) < 2 mg/liter and other clinically relevant thresholds. Characteristics of 78 patients receiving 600 mg/12 h of linezolid with a C(min) determination at the steady state and within the first 10 days of treatment were retrospectively reviewed. Concentrations were measured using high-pressure liquid chromatography. Univariate and multivariate analysis were performed to identify risk factors of low C(min). A total of 29.5% of patients had a C(min) < 2 mg/liter. The percentage was significantly higher in patients with an estimated glomerular filtration (eGF) > 80 ml/min, in intensive care unit (ICU) patients, and in patients with an infection due to Staphylococcus aureus. The independent predictors of C(min) < 2 mg/liter were an eGF > 80 ml/min (odds ratio [OR], 10; 95% confidence interval [CI], 2.732 to 37.037; P = 0.001) and infection due to S. aureus (OR, 5.906; 95% CI, 1.651 to 21.126; P = 0.006). A linezolid C(min) of <2 mg/liter was found in 29.5% of cases, and the risk was significantly higher among those with an eGF > 80 ml/min and in infections due to S. aureus. In patients with severe sepsis, a loading dose or continuous infusion and drug monitoring could improve the pharmacodynamic parameters associated with linezolid efficacy.
Sigma-Aldrich
Linezolid, ≥98% (HPLC)