Merck
PPARγ agonist from Chromolaena odorata.
Journal of natural products (2012-11-29)
Man-Li Zhang, Dianne Irwin, Xiao-Ning Li, Françoise Sauriol, Xiao-Wei Shi, Yu-Fang Wang, Chang-Hong Huo, Li-Geng Li, Yu-Cheng Gu, Qing-Wen Shi
ABSTRACT
A phytochemical investigation of Chromolaena odorata resulted in the isolation of five new compounds, 5aα,6,9,9aβ,10-pentahydro-10β-hydroxy-7-methylanthra[1,2-d][1,3]dioxol-5-one (1), 1,2-methylenedioxy-6-methylanthraquinone (2), 3-hydroxy-1,2,4-trimethoxy-6-methylanthraquinone (3), 3-hydroxy-1,2-dimethoxy-6-methylanthraquinone (4), and 7-methoxy-7-epi-medioresinol (5), together with 12 known compounds, odoratin (6), 3β-acetyloleanolic acid (7), ursolic acid (8), ombuin (9), 4,2'-dihydroxy-4',5',6'-trimethoxychalcone (10), (-)-pinoresinol (11), austrocortinin (12), tianshic acid (13), cleomiscosin D (14), (-)-medioresinol (15), (-)-syringaresinol (16), and cleomiscosin A (17). All the compounds were evaluated for their PPARγ transactivation activity, and compound 6 showed moderate activity with an EC(50) value of 3.10 μM.
Product Description
Supelco
Pinoresinol, analytical standard
Sigma-Aldrich
Pinoresinol, ≥95.0% (HPLC)
Sigma-Aldrich
2-Methylanthraquinone, technical, ≥95% (HPLC)