Merck
  • Proteomic response of Bacillus subtilis to lantibiotics reflects differences in interaction with the cytoplasmic membrane.
Proteomic response of Bacillus subtilis to lantibiotics reflects differences in interaction with the cytoplasmic membrane.
Antimicrobial agents and chemotherapy (2012-08-29)
Michaela Wenzel, Bastian Kohl, Daniela Münch, Nadja Raatschen, H Bauke Albada, Leendert Hamoen, Nils Metzler-Nolte, Hans-Georg Sahl, Julia E Bandow
ABSTRACT
Mersacidin, gallidermin, and nisin are lantibiotics, antimicrobial peptides containing lanthionine. They show potent antibacterial activity. All three interfere with cell wall biosynthesis by binding lipid II, but they display different levels of interaction with the cytoplasmic membrane. On one end of the spectrum, mersacidin interferes with cell wall biosynthesis by binding lipid II without integrating into bacterial membranes. On the other end of the spectrum, nisin readily integrates into membranes, where it forms large pores. It destroys the membrane potential and causes leakage of nutrients and ions. Gallidermin, in an intermediate position, also readily integrates into membranes. However, pore formation occurs only in some bacteria and depends on membrane composition. In this study, we investigated the impact of nisin, gallidermin, and mersacidin on cell wall integrity, membrane pore formation, and membrane depolarization in Bacillus subtilis. The impact of the lantibiotics on the cell envelope was correlated to the proteomic response they elicit in B. subtilis. By drawing on a proteomic response library, including other envelope-targeting antibiotics such as bacitracin, vancomycin, gramicidin S, or valinomycin, YtrE could be identified as the most reliable marker protein for interfering with membrane-bound steps of cell wall biosynthesis. NadE and PspA were identified as markers for antibiotics interacting with the cytoplasmic membrane.
Sigma-Aldrich
Nisin from Lactococcus lactis, potency: ≥900 IU/mg