Merck
  • Protective effect of diphenyl diselenide on ischemia and reperfusion-induced cerebral injury: involvement of oxidative stress and pro-inflammatory cytokines.
Protective effect of diphenyl diselenide on ischemia and reperfusion-induced cerebral injury: involvement of oxidative stress and pro-inflammatory cytokines.
Neurochemical research (2012-08-01)
César A Brüning, Marina Prigol, Cristiane Luchese, Cristiano R Jesse, Marta M M F Duarte, Silvane S Roman, Cristina W Nogueira
ABSTRACT
Cerebrovascular diseases, including ischemic stroke, are associated with high mortality worldwide. Oxidative stress and inflammation are important pathophysiological mechanisms involved in post-ischemic cerebral injury. The present study was designed to investigate the potential protective effect of diphenyl diselenide (PhSe)(2), an organoselenium compound with antioxidant and anti-inflammatory properties, against ischemia/reperfusion (I/R) insult in rat brain. The experimental model adopted was that of surgically-induced brain ischemia, performed by means of bilateral common carotid artery occlusion in rats. The effect of a single oral dose of (PhSe)(2) (50 mg/kg), administered 30 min before the onset of ischemia, was investigated by assessing cerebral oxidative stress-related biochemical parameters and pro-inflammatory cytokines in plasma of rats. The results demonstrated an increase in the levels of malondialdehyde (MDA), reactive oxygen species (ROS) and nitrate/nitrite as well as the alteration in the non-enzymatic and enzymatic (catalase and superoxide dismutase) antioxidant defense system induced by I/R insult in rat brain. I/R insult increased the levels of IL-1β, IL-6, TNF-α and INF-γ in plasma of rats. The administration of (PhSe)(2) restored cerebral levels of MDA, ROS, nitrate/nitrite and antioxidant defenses of rats exposed to I/R insult. (PhSe)(2) markedly reduced pro-inflammatory cytokines in plasma of I/R rats. I/R insult increased the plasma levels of tissue damage markers, such as creatine kinase and α-1-acid glycoprotein. Pretreatment with (PhSe)(2) was effective in reducing the levels of these proteins. In addition, (PhSe)(2) attenuated cerebral histological alterations induced by I/R. This study showed for the first time the in vivo protective effect of (PhSe)(2) against oxidative stress and pro-inflammatory cytokines-induced by I/R insult in rats.
Sigma-Aldrich
Diphenyl diselenide, purum, ≥97.0% (GC)
Sigma-Aldrich
Diphenyl diselenide, 98%