Merck
  • Perioperative use of etoricoxib reduces pain and opioid side-effects after total abdominal hysterectomy: a double-blind, randomized, placebo-controlled phase III study.
Perioperative use of etoricoxib reduces pain and opioid side-effects after total abdominal hysterectomy: a double-blind, randomized, placebo-controlled phase III study.
Current medical research and opinion (2012-06-29)
Eugene R Viscusi, Tara L Frenkl, Craig T Hartrick, Narinder Rawal, Henrik Kehlet, Dimitris Papanicolaou, Arnold Gammaitoni, Amy T Ko, Leslie M Morgan, Anish Mehta, Sean P Curtis, Paul M Peloso
ABSTRACT
To evaluate the effects of two different doses of etoricoxib delivered perioperatively compared with placebo and standard pain management on pain at rest, pain with mobilization, and use of additional morphine/opioids postoperatively. In this double-blind, placebo-controlled, randomized clinical trial, we evaluated postoperative pain following total abdominal hysterectomy over 5 days in patients receiving placebo or etoricoxib administered 90 min prior to surgery and continuing postoperatively. Patients were randomly assigned to receive either placebo (n = 144), etoricoxib 90 mg/day (n = 142), or etoricoxib 120 mg/day (n = 144). Average Pain Intensity at Rest over days 1-3 (0- to 10-point numerical rating scale [NRS]) was the primary efficacy endpoint. Secondary endpoints included Average Pain Intensity upon Sitting, Standing, and Walking over days 1-3 (0- to 10-point NRS) as well as Average Total Daily Dose of Morphine over days 1-3. This trial is registered on www.clinicaltrials.gov (NCT00788710). The least squares (LS) means (95% CI) for the primary endpoint were 3.26 (2.96, 3.55); 2.46 (2.16, 2.76); and 2.40 (2.11, 2.69) for placebo, etoricoxib 90 mg, and etoricoxib 120 mg, respectively, significantly different for both etoricoxib doses versus placebo (p < 0.001). Patients on etoricoxib 90 mg and 120 mg required ~30% less morphine per day than those on placebo (p < 0.001), which led to more rapid bowel recovery in the active treatment groups by ~10 hours vs. placebo. A greater proportion of patients on etoricoxib (10-30% greater than placebo) achieved mild levels of pain with movement, defined as pain ≤3/10. A key limitation for this study was that movement-evoked pain measurements were not designated as primary endpoints. In patients undergoing total abdominal hysterectomy, etoricoxib 90 mg and 120 mg dosed preoperatively and then continued postoperatively significantly reduces both resting and movement-related pain, as well as reduced opioid (morphine) consumption that led to more rapid bowel recovery.
Product Description
Supelco
Etoricoxib, VETRANAL®, analytical standard