Merck
  • L-pantoyl lactone dehydrogenase from Rhodococcus erythropolis: genetic analyses and application to the stereospecific oxidation of L-pantoyl lactone.
L-pantoyl lactone dehydrogenase from Rhodococcus erythropolis: genetic analyses and application to the stereospecific oxidation of L-pantoyl lactone.
Applied microbiology and biotechnology (2012-03-09)
Dayong Si, Nobuyuki Urano, Shinya Nozaki, Kohsuke Honda, Sakayu Shimizu, Michihiko Kataoka
ABSTRACT
The 1,2-propanediol (1,2-PD) inducible membrane-bound L-pantoyl lactone (L-PL) dehydrogenase (LPLDH) has been isolated from Rhodococcus erythropolis AKU2103 (Kataoka et al. in Eur J Biochem 204:799, 1992). Based on the N-terminal amino acid sequence of LPLDH and the highly conserved amino acid sequence in homology search results, the LPLDH gene (lpldh) was cloned. The gene consists of 1,179 bases and encodes a protein of 392 amino acid residues. The deduced amino acid sequence showed high similarity to the proteins of the FMN-dependent α-hydroxy acid dehydrogenase/oxidase family. The overexpression vector pKLPLDH containing lpldh with its upstream region (1,940 bp) was constructed and introduced into R. erythropolis AKU2103. The recombinant R. erythropolis AKU2103 harboring pKLPLDH showed six times higher LPLDH activity than the wild-type strain. Conversion of L-PL to ketopantoyl lactone was achieved with 92% or 80% conversion yield when the substrate concentration was 0.768 or 1.15 M, respectively. Stereoinversion of L-PL to D-PL was also carried out by using the combination of recombinant R. erythropolis AKU2103 harboring pKLPLDH and ketopantoic acid-reducing Escherichia coli.
Sigma-Aldrich
D-(−)-Pantolactone, 99%
Sigma-Aldrich
DL-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, purum, ≥97.0% (T)
Sigma-Aldrich
(S)-(+)-Pantolactone, 97%