Merck
  • Phototropin-dependent biased relocalization of cp-actin filaments can be induced even when chloroplast movement is inhibited.
Phototropin-dependent biased relocalization of cp-actin filaments can be induced even when chloroplast movement is inhibited.
Plant signaling & behavior (2011-11-08)
Noboru Yamada, Noriyuki Suetsugu, Masamitsu Wada, Akeo Kadota
ABSTRACT
In a recent publication using an actin-visualized line of Arabidopsis (Ichikawa et al. 2011, ref. 11), we reported a detailed analysis with higher time resolution on the dynamics of chloroplast actin filaments (cp-actin filaments) during chloroplast avoidance movement and demonstrated a good correlation between the biased configuration of cp-actin filaments and chloroplast movement. However, we could not conclusively determine whether the reorganization of cp-actin filaments into a biased configuration preceded actual chloroplast movement (and, thus, whether it could be a cause of the movement). In this report, we present clear evidence that the reorganization of cp-actin filaments into a biased distribution is induced even in the absence of the actual movement of chloroplasts. When the cells were treated with 2,3-butanedione monoxime (BDM), a potent inhibitor of myosin ATPase, chloroplast motility was completely suppressed. Nevertheless, the disappearance and biased relocalization of cp-actin filaments toward the side of the prospective movement direction were induced by irradiation with a strong blue light microbeam. The results definitively indicate that the reorganization of cp-actin filaments is not an effect of chloroplast movement; however, it is feasible that the biased localization of cp-actin filaments is an event leading to chloroplast movement.
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%
Supelco
2,3-Butanedione monoxime, for spectrophotometric det. of urea, ≥99.0%